Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Heidolph™ Radleys PTFE Anchor Stirrer Paddle
PTFE Stirring Paddle, for use with Radleys Carousel Reaction Workstations
Supplier: Heidolph™ 015861820
Description
Anchor PTFE Stir Paddle for 250mL FlasksSpecifications
PTFE | |
Anchor Paddle | |
250 mL Flasks |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction