Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Heidolph™ Separatory Funnel Attachment for Platform Shaker
For use with Unimax 1010, Duomax 1030, Polymax 1040, Promax 1020 platform shakers
Supplier: Heidolph™ 036303340
Description
Attachment for 4 separatory funnels (50mL, conical) or 4 separatory funnels (100mL, conical)Specifications
Separatory funnel attachment | |
PharMed™ Two-Stop | |
0.2 cm | |
For Unimax 1010, Duomax 1030, Polymax 1040, Promax 1020 Platform Shakers |
Safety and Handling
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction