Learn More
Heidolph™ Platform Shaker Accessories
SDP
Separatory funnel clamps for Heidolph Platform Shakers as indicated
Supplier:  Heidolph™ 036303460
 View more versions of this product
Catalog No. 13-889-846
Add to Cart
Description
Description
  • Choice of clamp either for volumes 250, 500, and 1000 mL or 2000 mL separatory funnel
Specifications
Specifications
2 L
Separatory funnel clamp
Polymax 2040, Promax 2020, Unimax 2010 platform shakers
Product Suggestions
Product Suggestions
Safety and Handling
Safety and Handling
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov