Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Heidolph™ Lara Controlled Lab Reactor Accessory, Probe Adapter
Temperature probes
Specifications
Type | Adapter |
---|---|
Material | PTFE |
For Use With (Equipment) | Lara controlled lab reactor |
Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity & Availability | |||||
05-406-431
|
Heidolph™
015900260 |
100mL | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||||
05-406-463
|
Heidolph™
015907069 |
1000, 2000, 5000 and 10000mL | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||||
Specifications
Adapter | |
PTFE | |
Lara controlled lab reactor |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title