Learn More
Heidolph™ Rotary Evaporator Accessories - Interchangeable Glassware
SDP
Five condenser assemblies allow customization of evaporator glassware for any application
Supplier:  Heidolph™ 036301880
 View more versions of this product
Catalog No. 13-878-645
Add to Cart
Description
Description
  • All glassware can be interchanged among rotary evaporator models
  • Each set includes one 1000 mL evaporating flask and one 1000 mL receiving flask
  • All sets are available with standard or plastic safety-coated glassware (halar coating for G5B and abcite coating for G1B/G3B/G6B)
  • Evaporating flasks and vapor tubes with the standard ground joint NS 24/40 are also available with NS 29/32 ground joints
Specifications
Specifications
G6 Vertical Condenser
Rotary Evaporator
Valve
Product Suggestions
Product Suggestions
Safety and Handling
Safety and Handling
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov