Learn More
Heidolph™ Hei-Vap Rotary Evaporator Temperature Sensors
Accessory parts and components for Heidolph Hei-VAP Rotary Evaporators
$682.50 - $828.00
Catalog Number | Mfr. No. | Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Type | Price | Quantity & Availability | |||||
13-889-340
|
Heidolph™
036308010 |
Vapor Temperature Sensor |
Each for $828.00
|
|
|||||
13-889-341
|
Heidolph™
036308020 |
AUTOaccurate Sensor |
Each for $682.50
|
|
|||||
Description
AUTOaccurate-sensor
- For Hei-VAP Precision, Hei-VAP Expert Control, and Hei-VAP Ultimate Control
- Only in combination with glassware set G3 and G6
- For Hei-VAP Advantage Hei-VAP Precision, Hei-VAP Expert, Hei-VAP Ultimate, Hei-VAP Expert Control, and Hei-VAP Ultimate Control
Specifications
Hei-VAP rotary evaporator |