Learn More
Heidolph™ Flask Holders for Platform Shakers
Accessory for Duomax 1030, Polymax 1040, Promax 1020, and Unimax 1010 platform shakers
Supplier: Heidolph™ 036303270
Description
Holds Erlenmeyer flasks securely.
Specifications
50 mL | |
Flask Holder | |
Heidolph Duomax 1030, Polymax 1040, Promax 1020 and Unimax 1010 Platform Shakers | |
14 |
Safety and Handling
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov