Learn More
Sklar Instruments Hegar Uterine Dilator, Sklar, Double End 3mm/4mm, Curved, Stainless Steel. EA/1.
Supplier: Sklar Instruments 904804
Hegar Uterine Dilator, Sklar, Double End 3mm/4mm, Curved, Stainless Steel. Sklar's Hegar Double-Ended Uterine Dilator is a manual, probe-style instrument used to dilate the cervix. They are solid, nonflexible, and available in multiple diameters. These dilators are slightly curved in opposite directions from the center and have smooth, rounded tips. This product is the 3mm/4mm dilator. The overall length is 8 inches.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.