Learn More
OHAUS™ Heavy Duty Shaker
OHAUS Heavy Duty Shakers are designed to handle a range of applications. Available as analog or digital model with microprocessor control to provide variable speed and consistent shaking, while ramping to the set speed
Supplier: OHAUS™ 30391842/EMD
Description
- Patented Accu-Drive Shaking System
- LED displays for speed and time
- Built-in tray and non-slip rubber mat included.
- Overload protection and speed ramping feature
- Calibration mode for speed
Specifications
25 mm | |
110 V | |
25.4 cm | |
88.9 cm | |
18 in. | |
45.7 cm | |
SHHD2325DG |
20 to 500 rpm | |
50 lb. | |
24 in. | |
61 cm | |
84.5 cm | |
Digital | |
Non-skid rubber mat, 3 wire cord and plug |