Learn More
LUMENITE CONTROL TECHNOLOGY HEAVY DUTY SANITROD W 1-1/2IN
Supplier:  LUMENITE CONTROL TECHNOLOGY HDDCFT1.5IN6IN
Encompass
heavy duty sanitrode with 1-1/2in tri-clamp tank connection with 6in long Teflon insulated probe
Catalog No. NC2091010