Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Vikan Heavy-Duty Push Broom, 24 in.
Designed from the US food and beverage manufacturer feedback, the Heavy-Duty Push Broom has key features that those with large areas to clean will love.
$40.88
Specifications
Product Type | Broom Head |
---|---|
Sterility | Non-sterile |
Autoclavable | Autoclavable |
Color-coded | Yes |
Material | Polypropylene, Stainless Steel |
Catalog Number | Mfr. No. | Color Code | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Color Code | Price | Quantity & Availability | |||||
17-467-551
|
Vikan
31959 |
Black |
Each for $40.88
|
|
|||||
17-467-552
|
Vikan
319588 |
Gray |
Each for $40.88
|
|
|||||
17-467-553
|
Vikan
31958 |
Purple |
Each for $40.88
|
|
|||||
17-467-554
|
Vikan
31957 |
Orange |
Each for $40.88
|
|
|||||
17-467-555
|
Vikan
319566 |
Brown |
Each for $40.88
|
|
|||||
17-467-556
|
Vikan
31956 |
Yellow |
Each for $40.88
|
|
|||||
17-467-557
|
Vikan
31955 |
White |
Each for $40.88
|
|
|||||
17-467-558
|
Vikan
31954 |
Red |
Each for $40.88
|
|
|||||
17-467-559
|
Vikan
31953 |
Blue |
Each for $40.88
|
|
|||||
17-467-560
|
Vikan
31952 |
Green |
Each for $40.88
|
|
|||||
Description
- Heavier Broom Head to maintain contact.
- Angle cut bristles helps to prevent skipping or hopping.
- Flared bristles for hard to reach areass.
- Densely packed stiff and soft bristles to sweep both and fine and larger debris.
Specifications
Broom Head | |
Autoclavable | |
Polypropylene, Stainless Steel |
Non-sterile | |
Yes |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title