Learn More
Glas Col LLC Heating Mantle
Supplier: Glas Col LLC 100A MO1063
Fabric heating top covers the top of the flask to reduce vapor condensation during distillation, also protects the flask. For 500ml 1-2 or 3 neck flasks heats to 400 degrees C maximum internal operating temperature. 115V, 140W, CSA.