Learn More
LabStrong Corp HEATER SHEET
Supplier: LabStrong Corp 562X24LS
LabStrong part number 562X24LS compares to Fi-Streem part number 562X24. The heating element sheath is a replacement part for Fi-Streem Water Purification Stills. The heater sheath comes with the plastic element union.