Learn More
Braintree Scientific Heat Lamp with table clamp
SDP
Supplier:  Braintree Scientific HL1120V
Encompass
Heat Lamp, HL-1 Arm Length: 2 spring loaded arms each 18 lengths Table Clamp : Standard U screw clamp expands to fit bench up to 2.25 deep Weight: 2 lbs. Power: 110VAC Bulb: 250 watt infrared heat Lamp Infrared heat lamp is table mounted with an adjustable swing arm providing for easy adjustment of radiant heat to specimen. The lamp is supplied with a 250W(infrared) heat lamp bulb and a built in reflector. A chrome bulb guard is also provided and easily snaps onto the lamp housing after the bulb is installed. Fits standard 110V AC socket. **Sold for research purposes only and not for human use**
Catalog No. 50-195-4936
Due to product restrictions, please to purchase or view availability for this product.