Learn More
Ace Glass, Inc. Heat gun, Master-Mite; 10008, 650°F, 3.8CFM, 1.75lbs, cULus rated
Supplier: Ace Glass, Inc. 1207220
Economical, compact, flameless heat gun for heat shrinkage as well as countless other industrial applications. Input is 120v, 5.4 amps. Supplied with 2.1m, three-conductor grounded cord. Unit consists of heat gun with silver nozzle and deflector adapter. CULus Listed