Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
binx™ health io™ Male Urine Specimen Collection Kit
The Male Urine Specimen Collection Kit is required for the collection, transportation, and preservation of DNA in a male urine sample for use with the binx™ health io™ CT/NG Assay.
Supplier: binx™ 5.002.001
Description
The binx™ health io™ Male Urine Specimen Collection Kit is required for the collection, transportation, and preservation of DNA in a male urine sample for use with the binx™ health io™ CT/NG Assay.
- Kit consists of a screw cap tube containing Transport and Preservation Medium and a Transfer Pipet
- Specimens stored and transported using the binx™ health io™ Male Urine Specimen Collection Kit are intended to be processed using the binx health io CT/NG Assay using the binx™ health io™ Instrument
- It is not intended to be used for the preservation and transfer of samples for any other test
- Patient samples may be stored in the Collection Kit for 24 hours at room temperature or for seven (7) days refrigerated at 2°C to 8°C
Specifications
Waived | |
Male Urine | |
Collection Kit | |
Liquid |
Chlamydia trachomatis, N. gonorrhoeae | |
High | |
Chlamydia Testing |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction