Learn More
Molecular Dimensions HC35 LIQUID NITROGEN DEWAR
Supplier: Molecular Dimensions MD744
HC35 - High Capacity Crystal Storage Dewar designed for storing large quantities of a variety of materials at cryogenic temperatures.Large storage capacity - 850 cryovials (5/cane)- 1020 (6/cane). Holds 35 L of liquid Nitrogen.