Learn More
STEMCELL Technologies HBV (HLA Class I Control) Peptide Pool, Size: 1 Unit
SDP
Supplier:  STEMCELL Technologies 1000673
Encompass_Preferred
HBV (HLA Class I Control) Peptide Pool is a lyophilized mixture of 9 peptides from the hepatitis B virus (HBV). The pool consists of defined HLA class I-restricted T cell epitopes and can be used as an antigen-specific positive control to stimulate T cells.
Catalog No. 50-239-0221