Learn More
Apexbio Technology LLC HBTU, 50g. Cas: 94790-37-1 MFCD: MFCD27957357. Peptide coupling reagent
SDP
Supplier:  Apexbio Technology LLC A702350000
Encompass_Preferred
IC50: Not available.HBTU (2-(1H-benzotriazol-1-yl)-1,1,3,3-tetramethyluronium hexauorophosphate), a coupling reagent commonly used in solid phase peptide synthesis. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4968
Due to product restrictions, please to purchase or view availability for this product.