Learn More
Nova One Diagnostics HBIC LINIARITY TEST
Supplier: Nova One Diagnostics HBLG04041100
Liquid HbA1c Linearity Combo kit, 180 day refrigerated stability, 1 mL L1 + 1 mL L2 + 1mL L3 + 1 mL L4 (4 ml Kit) plus 100 disposable pipettes