Learn More
Perkin Elmer US LLC HATR Trough Top Plate - ZnSe, 45°
Supplier: Perkin Elmer US LLC L1272338
Based on the Attenuated Total Reflectance (ATR) principle, the PerkinElmer Horizontal ATR (HATR) Accessory offers a fast, reliable means for analyzing a wide range of solid, liquid, and paste samples not easily examined by conventional transmission techniques.