Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Harris Industries Colored Delineator Tape
Designed for safety marking applications in industrial plants, commercial buildings, institutions and recreational facilities.
Supplier: Harris Industries HT208BY
Description
Features:
- Alternating color stripes
- Highly visible
- Pressure sensitive
- Rubber-based permanent adhesive
- Self-wound
- Clear laminate resists oil, dirt, chemicals and abrasion
- 6.0mil/ 0.150mm thickness
- Laminated PVC will not fade or scuff
- Holds up to traffic and wear longer than paint
- Can be applied quickly and easily with minimal surface preparation
- Maintenance-free; brilliance is restored with wet mopping/washing
Red/White: Fire Protection Equipment; Black/Yellow: Physical Hazard; Black/White: Housekeeping Aisle Marking
Compliance
OSHA 1910.144
Specifications
14°F to 176°F | |
3 in. | |
-10°C to +80°C | |
36 yd. x 3 in. (32.9 m x 7.62 cm) | |
32.9 m | |
7.62 cm (3 in.) | |
OSHA 1910.144 | |
Black, Yellow |
23N/cm | |
7.62 cm | |
6 mil | |
PVC | |
36 yd. | |
Rubber-based Adhesive | |
Safety marking applications in industrial plants, commercial buildings, institutions and recreational facilities, resistance to oil, dirt and chemicals | |
Hazard Warning Tape |
Safety and Handling
ShelfLife : 1 Year
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction