Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Harris Industries Colored Delineator Tape

Designed for safety marking applications in industrial plants, commercial buildings, institutions and recreational facilities.

Supplier:  Harris Industries HT200B

 View more versions of this product

Catalog No. 19-019996


Add to Cart

Description

Description

Features:

  • Alternating color stripes
  • Highly visible
  • Pressure sensitive
  • Rubber-based permanent adhesive
  • Self-wound
  • Clear laminate resists oil, dirt, chemicals and abrasion
  • 6.0mil/ 0.150mm thickness
Benefits:
  • Laminated PVC will not fade or scuff
  • Holds up to traffic and wear longer than paint
  • Can be applied quickly and easily with minimal surface preparation
  • Maintenance-free; brilliance is restored with wet mopping/washing

Red/White: Fire Protection Equipment; Black/Yellow: Physical Hazard; Black/White: Housekeeping Aisle Marking

Compliance

OSHA 1910.144

Specifications

Specifications

Hazard Warning Tape
18 yd. x 2 in. (16.4 m x 5.08 cm)
Rubber-based Adhesive
2 in.
23N/cm
-10°C to +80°C
18 yd.
14°F to 176°F
PVC
Black, White
OSHA 1910.144
5.08 cm
6 mil
Safety marking applications in industrial plants, commercial buildings, institutions and recreational facilities, resistance to oil, dirt and chemicals
16.4 m
Product Suggestions

Product Suggestions

Safety and Handling

Safety and Handling

ShelfLife : 1 Year

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit