Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Harris Industries Barricade Tapes

Increase visibility of hazards to help prevent accidents

Supplier:  Harris Industries BT20

 View more versions of this product

Catalog No. 19-019956


Add to Cart

Description

Description

Strong, lightweight polyethylene tapes with continuous repetition of message.

  • Can be tied, stapled, or nailed into place
  • Economical and reusable
  • “ Caution” tapes have yellow background
  • “Danger” tapes have red background

Order Info

Roll size: 3 in. W x 1000 ft.L (7.6cm x 304.8m). Thickness: 4mil.

Compliance

Meet OSHA 1910.144 for marking physical hazards.

Specifications

Specifications

Caution Tape
1000 ft. x 3 in. (304.8 m x 7.62 cm)
3 in.
304.8 m
Increase visibility of hazards to help prevent accidents
Magenta, Yellow
HDPE
7.62 cm
4 mil
OSHA 1910.144
1000 ft.
Caution People Working
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit