Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Hardy Diagnostics™ Coagulase Cyro™

The ready-to-use Coagulase Cryo is used to differentiate Staphylococcus species based on the ability to produce coagulase
Supplier: Hardy Diagnostics™ Z202
This item is not returnable.
View return policy
Description
- Frozen rabbit plasma
- Tube or slide coagulase test
- Staphylococcus aureus detection
- 0.5mL in 2mL vials
- Ready to use
Certifications
ISO 13485
Specifications
-2°C | |
Staphylococcus Coagulase Test | |
Vials (0.5mL in 2mL vials) |
Liquid | |
Used for the identification of Staphylococcus aureus | |
20 Pack |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title