Learn More
HANNA INSTRUMENTS INC Ammonia High Range Photometer
SDP
Supplier:  HANNA INSTRUMENTS INC HI97733C
Encompass
The HI97733 Ammonia Photometer combines accuracy and ease of use in a simple, portable design. The advanced optical system provides lab-quality accuracy while its user-friendly design is easy for any user making it the perfect photometer for your water quality testing needs. The HI97733 measures ammonia in water samples from 0.0 to 100.0 mg/L (ppm).No warm up time needed before taking a measurement.Tutorial mode for easy step-by-step instructions.Displays results in multiple chem
Catalog No. NC1890594