Learn More
Cole-Parmer Oakton EcoTestr™ pH 2+ Pocket pH Meter
Supplier: Cole-Parmer 3542301
Additional key provides an easier, more intuitive interface. Improved cap functions as a sample cup and as a stabilizing base to stand the meter upright. IP67-rated housing is waterproof and floats. This new addition to the Oakton EcoTestr product line features updates to make measuring water quality easier and faster. The display is larger with a better viewing angle, has indicators for battery life, readiness, and calibration, and shows both the parameter and temperature readings at the same time. To enhance field use, the cap was redesigned to be leakproof and can be attached to the top of the meter when not being usedno more lost caps! The cap features a fill line, so you know how much sample you need for an accurate reading when using the cap as a sample cup. It is also wider, providing a base to keep the meter upright for hands-free measurements. The keypad now has a fourth key to simplify menu navigation while keeping convenient one-touch calibration.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.