Learn More
MilliporeSigma™ Supelco™ Hamilton™ Needles for RN Syringes, Blunt Tip, Stainless Steel
Hamilton™ needles for RN syringes are bevel tip needles, designed for optimum septum penetration and to prevent septum coring.
Supplier: MilliporeSigma™ Supelco™ 19141U
Specifications
Stainless Steel (Tip) | |
2 in. | |
6/Pk. | |
51 mm |
26 G | |
Blunt Tip | |
250 μL to 10 mL | |
Hamilton RN Syringes |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Hamilton is a registered trademark of Hamilton Co.