Learn More
MilliporeSigma™ Supelco™ Hamilton™ Needles for RN Syringes, Bevel Tip, Stainless Steel
Hamilton™ needles for RN syringes are bevel tip needles, designed for optimum septum penetration and to prevent septum coring.
Supplier: MilliporeSigma™ Supelco™ Z1181256EA
Specifications
Stainless Steel (Tip) | |
4 in. | |
6/Pk. | |
101.5 mm |
22 G | |
Bevel Tip | |
2.5 to 100 μL | |
Hamilton RN Syringes |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Hamilton is a registered trademark of Hamilton Co.