Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Supelco™ Hamilton™ Needles for LT and TLL Syringes, KEL-F hub (KF)
Supplier: MilliporeSigma™ Supelco™ 20756
Specifications
Stainless Steel (Tip) | |
28 G | |
2 in. | |
6/Pk. | |
2 |
Has KEL-F hub (KF) | |
KEL-F hub (KF) | |
Bevel Tip | |
51 mm | |
Hamilton LT and TLL Syringes |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Hamilton is a registered trademark of Hamilton Co.
Spot an opportunity for improvement?
Provide Content Correction