Learn More
Hamilton™ 900 Series Syringe, Sold by MilliporeSigma™ Supelco™
The Economical Version of Hamilton™ 800 Series Syringes
Supplier: MilliporeSigma Supelco 20906U
Specifications
Not Autoclavable | |
26s G | |
0.005 in. | |
0.019 in. | |
0.343 mm | |
0.483 mm | |
Microliter™ Syringe | |
122°F | |
50°F | |
2 | |
1 Ea. | |
Hamilton | |
Bevel |
Borosilicate Glass (Barrel), Stainless Steel (Tip) | |
0.014 in. | |
0.13 mm | |
0.260 in. | |
6.60 mm | |
2 in. | |
2000 psig (137.9 bar) | |
50°C | |
95 N | |
10°C | |
5 μL | |
51 mm | |
Cemented |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Microliter is a trademark of Hamilton Co.