Learn More
Hamilton™ 1000 Series Syringe, Removable Needle (RN), Sold by MilliporeSigma™ Supelco™
Gas-tight design minimizes the gas exchange in GC analysis. Syringes provide accurate volume delivery with a plunger that is mostly unsurpassed.
Supplier:  MilliporeSigma Supelco 22194U
 View more versions of this product
Catalog No. 11-101-0746
Due to product restrictions, please to purchase or view availability for this product.
Description
Description
Specifications
Specifications
Autoclavable
22 G
0.0161 in.
0.0283 in.
10.3 mm
0.72 mm
Gastight™ Syringe
239°F
50°F
2
1 Ea.
Hamilton
Bevel
Stainless Steel (Tip)
0.406 in.
0.41 mm
0.530 in.
13.5 mm
2 in.
200 psig (13.8 bar)
115°C
1005 RN
10°C
5 mL
51 mm
Removable
Product Suggestions
Product Suggestions
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
GASTIGHT is a registered trademark of Hamilton Co.