Learn More
SCANTIBODIES LABORATORY INC HAMA POSITIVE HUMAN PLASMA (3PH490)
Supplier: SCANTIBODIES LABORATORY INC 3PH490
The 3PH490 HAMA Positive Human Plasma from Scantibodies Laboratory, Inc. is composed of plasma from human origin with a HAMA value of 459.190 mg/mL. Its physical and chemical properties are as a slightly turbid, yellow to brown colored frozen liquid, with common plasma odor.