Learn More
Enzo Life Sciences Hydrogen peroxide fluorometric detection kit (500tests)
SDP
Supplier:  Enzo Life Sciences ADI907028
Encompass_Preferred
The Hydrogen Peroxide fluorometric detection kit provides fast results in just 10 minutes for the measurement of either hydrogen peroxide or peroxidase activity in a convenient one-step homogeneous assay. Sensitivity: (range 0 - 10 µM (H2O2); 0 - 25 mU/ml (peroxidase). Applications: Fluorescent detection, HTS . Species reactivity: Species independent .
Catalog No. 50-200-7086