Learn More
Ace Glass, Inc. Extraction apparatus, liquid/liquid, complete with all components to perform lighter than and heavier than water solvent extraction
Supplier: Ace Glass, Inc. 684650
Complete liquid-liquid extraction apparatus for use with lighter or heavier than water extracting solvents. Overflow and boil-up tube uses Ace-Thred connections allowing easy removal for cleaning, reducing cross-contamination among samples.