Learn More
H-B Instrument™ Durac™ Liquid-in-Glass Oven Thermometers
Total immersion type without featuring swell
Supplier:  H-B Instrument™ B608030900
 View more versions of this product
Catalog No. 13-201-959
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
View Alternate Product
Description
Description
  • Contains red organic liquid against white back glass
  • Individually packaged in recyclable packaging with a non-roll fitting
  • Individually serialized
Specifications
Specifications
Total Immersion
135 mm
Organic
20°C
±2°C
5.3 in.
Glass
130°C
20°C to 130°C
2°C
Product Suggestions
Product Suggestions