Learn More
H-B Instrument™ Enviro-Safe™ General Purpose Liquid-In-Glass Thermometers, Total Immersion
SCI_ED
Supplier:  H-B Instrument™ B605060000/EMD
 View more versions of this product
Catalog No. S43368
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Alert:
Individually serialized thermometers are traceable to NIST; include a multi-language Statement of Accuracy and Traceability.
Description
Description
Certified environmentally safe thermometers feature green, non-toxic, biodegradable, Enviro-Safe™ liquid against lead-free, white back glass.
  • Earth friendly, EnviroKleen™ certified thermometers
  • Meet government, education, and industry mandates to purchase “Green” products
Applications:
Ideal for use in chemical/petrochemical laboratories, universities, scientific institutions, food and beverage processing, pharmaceutical and medical industries, dairies, breweries and distilleries.
Order Info
Should you require an individualized Thermometer Calibration Report
Certifications
Manufactured and tested by USA quality team.
Specifications
Specifications
Individually serialized thermometers are traceable to NIST; include a multi-language Statement of Accuracy and Traceability.
Total Immersion
300 mm
50°C
±1°C
Ideal for use in chemical/petrochemical laboratories, universities, scientific institutions, food and beverage processing, pharmaceutical and medical industries, dairies, breweries and distilleries.
12 in.
Total Immersion Thermometer
-35°C
-35°C to +50°C
1°C
Product Suggestions
Product Suggestions