Learn More
H-B Instrument™ Enviro-Safe™ Environmentally Friendly Liquid-In-Glass Pocket Thermometers
Supplier: H-B Instrument™ B605700800
Alert:
Manufactured and tested by USA quality team and include a multi-language (English, French, German, Italian, Portuguese, and Spanish) Statement of Accuracy and Traceability.
Description
These EnviroKleen™ certified thermometers contain nontoxic, biodegradable Enviro-Safe liquid and are housed in a protective case for ‘on-the-go’ use and safety.
EnviroKleen™ certified thermometers feature green, nontoxic, biodegradable, Enviro-Safe liquid against lead-free, white back glass. Total immersion pocket thermometers are a compact 160mm (6.3 in.) long. Come with a case to protect from breakage.Case options include:
- Plastic Window - encased in plastic with an open window
- Metal Window - encased in metal with an open window
- Closed Plastic - housed in removable plastic case
- Closed Metal - housed in a removable metal case
- Aluminum Duplex - encased in nickel-plated brass with an open window and housed in a removable aluminum case
- Nickel-Plated Brass Window - encased in nickel-plated brass with an open window
Order Info
Should you require an individualized Thermometer Calibration Report
Specifications
Traceable to NIST. Includes a multi-language (English, French, German, Italian, Portuguese, and Spanish) Statement of Accuracy and Traceability. | |
Total Immersion | |
6.3 in. | |
120°F | |
20°F to 120°F | |
±1°F |
Ideal for field studies, waste water treatment and any “on-the-go” applications. | |
Closed plastic case | |
160 mm | |
20°F | |
— | |
1°F |