Learn More
Gyro Studies Lab Stool Kit
Unique lab stool has a turntable built into the seat.
Supplier: Science First 18725
Description
LabStool
- Made from interlocking parts, stool can be disassembled for easy storage
- Includes glue for permanent assembly
- Seat is slotted to accept rotational accessories; included insert allows user to develop personalized attachments
- Gyro Studies Kit
- Lab stool with bicycle wheel gyroscope, two accelerometers, one rotary mounting arm and two bag toss masses
Specifications
Lab Stool | |
For gyro studies | |
Glue for permanent assembly |