Learn More
GVS PCTE Filter Membranes: Plain Hydrophilic
Smooth, flat surface for even distribution in a single plane.
Supplier: GVS 1215620
Description
Smooth, flat surface for even distribution in a single plane.
Polycarbonate Track-Etched membranes are manufactured under Class 100 conditions to be exceptionally contaminant free. Exhibit negligible adsorption/absorption of filtrate, precise pore size, narrow pore distribution, low tare weights, low trace element level.
- Can be made hydrophobic
- Created with pore density and size precisely controlled
- Biologically inert
- Optically translucent (actually transparent in most pore sizes)
- Not stained by most solutions
- Nonhygroscopic
- Superior strength, chemical resistance, thermal stability
- Autoclavable
- Tare weight: 1.0mg/cm2
- Tensile strength: >3000psi
- Residual ash: 0.92mg/cm2
- Specific gravity: 0.94 to 0.97
- Minimum burst strength: 10psi
- Max. use temperature: 140°C (284°F)
Applications:
Recommended for viewing sample on filter surface: observation of sample with optical or electron microscopes; cold sterilization, tissue culture, fractionation, blood studies, virology, general separations.
Order Info
- Gamma irradiation or EtO
- Low extractables
Compliance
Pass USP Class VI Testing
Specifications
7.5L/min./cm2 | |
9psi | |
Polycarbonate Track-Etched (PCTE) | |
Sterile | |
General filtration, removal of red blood cells from plasma, flow control of reagents through assay, precise filtration and prefiltration | |
5μg/cm2 | |
284°F | |
207 bar | |
1.8 in. |
Autoclavable | |
Microporous Filter | |
0.6 μm | |
60mL/min./cm2 | |
Hydrophilic | |
Smooth, flat | |
140°C | |
5 to 12 μm | |
47 mm |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.