Learn More
Electron Microscopy Sciences Gusto High Speed Mini Centrifuge
Supplier: Electron Microscopy Sciences 7235912
This compact centrifuge is ideal for most protocols calling for fast spins (up to 12,500 rpm / 9,800 rcf) and quick spindowns. Useful for PCR, microfilter cell separation, clinical applications, and HPLC protocols. Operation is extremely simple for this high-level of sophistication. For duplicate spins, simply reload rotor and push Start. Designed with quick-spin option for commonly used 20-second spins. Includes a toolfree, quick-change rotor system. Safety features ensure the unit will not operate with an open lid, a jammed rotor or when out of balance. Efficiently designed for minimum bench top space.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.