Learn More
Electron Microscopy Sciences Forensic Gunshot Residue Field Kit with 5 Disc
Supplier: Electron Microscopy Sciences 7648015
Our comprehensive line of high-quality GSR field sampling kits includes certified SEM pin stubs with double-coated adhesive carbon tabs in plastic tubes. The certified SEM pin stubs have very low amounts of Pb, Sb and Ba, not detectable by SEM/EDX. The caps securely hold the GSR sample disc for easy GSR collection. Pre-labeled discs and tubes are both ready-for-sampling. A sturdy, sealed tamper evident transport box is also included. Compatible with FBI requirements, with four discs plus a control disc.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.