Learn More
Innovative Research Inc Guinea Pig Red Blood Cells 10ml
Supplier:  Innovative Research Inc IGPRBC10ML
Encompass_Preferred
Guinea Pig Red Blood Cells from Innovative Research are manufactured using whole blood collections from Guinea Pigs located in the USA. The whole blood is washed to remove the platelet-rich plasma, buffy coat layer, and leukocytes. Red blood cells are perishable and are collected and processed upon receipt of order. We offer reliable, consistent products that deliver reliable, consistent results.
Catalog No. 50-203-4851