Learn More
Colorado Serum Co GUINEA PIG ALSEVERS 15ML BLD
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Colorado Serum Co 30100
![Encompass](/content/dam/fishersci/glyphs/encompass.png)
GUINEA PIG ALSEVERS 15ML BLOOD. Blood containing alsevers solution is a 50% mix of whole blood and alsevers solution. Alsevers solution, which is made in our laboratory and tested for sterility, acts as an anti-coagulant and preservative. It contains sodium citrate and glucose.