Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
OHAUS™ Guardian™ 2000 Hotplate
Guardian™ 2000 Hotplate is ideal for education and other lab environments that require affordable, everyday fast heating to 15 L. All three sizes offer simple knob control to easily adjust the heating functions.
Supplier: OHAUS™ 30680260/EMD
This item is not returnable.
View return policy
Description
- Applications: Boiling Liquids, Basic Sample and Solution Preparation, Dissolving Buffers and Reagents, Preparing Media, Concentrating Samples
- Display: Prominent hot top indicator light, green indicator lights when unit is heating and stirring
- Operation: Simple knob control, stirring up to 1600 rpm, temperature up to 500°C (ceramic top) or 380°C (ceramic coated stainless steel top), power cord (included), stirring models include PTFE coated stir bar
- Communication: N/A
- Construction: Durable metal housing, PET front panel, control knobs, ON/OFF switch for heater disconnect, built in support rod holder, easy to clean resin, ceramic or ceramic coated stainless steel tops
- Design Features: Hot top indicator light to protect user, green indicator lights illuminate when unit is heating and stirring
Specifications
264 x 163 x 101 mm (10.4 x 6.4 x 4 in.) | |
Round | |
Porcelain-coated Stainless Steel | |
Analog | |
50°C to 380°C | |
2.5 kg | |
5 A | |
e-G21HPRDS |
15 L (4 gal.) | |
100/120 V | |
122°F to 716°F | |
135 mm dia. | |
5.5 lb. | |
505 W | |
50/60 Hz |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction