Learn More
World Precision Instrument GSNO S-nitrosoglutatione-GSNO-100
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: World Precision Instrument GSNO100
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
GSNO has been identified in vivo as a potential storage and transport vehicle for NO in the body. GSNO has been used in clinical trials to treat a form of preeclampsia and to prevent platelet aggregation. It also has considerable potential as NO donor in medicine.