Learn More
World Precision Instrument GSNO S-nitrosoglutatione-GSNO-50
Supplier: World Precision Instrument GSNO50
GSNO has been identified in vivo as a potential storage and transport vehicle for NO in the body. GSNO has been used in clinical trials to treat a form of preeclampsia and to prevent platelet aggregation. It also has considerable potential as NO donor in medicine.