Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GSC Go Science Crazy Electroscopes Flask Type
SCI_ED

Visual demonstration of electrostatic forces.

Supplier:  GSC Go Science Crazy 450150

Catalog No. S43262A


Add to Cart

Includes: Instruction sheet

Description

Description

  • Consists of aluminum leaves that are mounted on a metal rod
  • Rubber stopper holds rod inside the 250mL Borosilicate Erlenmeyer flask
  • Size 3.5 x 3.5 x 6.5 in., weight 0.5 lb.

Specifications

Specifications

Instruction sheet
Flask Type Electroscope
0.22 kg
0.5 lb.
Product Suggestions

Product Suggestions

Safety and Handling

Safety and Handling

WARNING: Cancer - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit