Learn More
GSC Go Science Crazy Glass Burette with PTFE Stopcock
Avoid hassles by choosing a burette with a stopcock that won't freeze. The GSC International Glass Burette with PTFE Stopcock is subdivided by 0.1 ml, does not require grease and is nearly freeze proof.
$30.30 - $140.90
Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity & Availability | |||||
S04822A
![]() |
GSC Go Science Crazy
928025 |
50 mL |
Each for $30.30
Pack of 5 EA for $137.60 |
|
S04882B
![]() |
GSC Go Science Crazy
928035 |
100 mL |
Pack of 5 for $140.90
|
|
S04822
![]() |
GSC Go Science Crazy
928015 |
25 mL | This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
Description
- The PTFE stopcock is almost freeze proof and does not require grease
- Subdivided by 0.1 ml
- Made from borosilicate glass
Specifications
Burette | |
Glass |
Not a toy. Product intended for ages 13 and up.