Learn More
Electron Microscopy Sciences TEM Window Pure Silicon, (2) 100x1500 µm, 15nm
Supplier: Electron Microscopy Sciences 7604277
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Pure Silicon TEM Windows feature imaging windows with 5 to 15 nm thickness, reducing background contribution and interference for higher contrast imaging. The membrane can be vigorously plasma cleaned to remove organic contamination, unlike traditional carbon grids. The elemental silicon composition of TEM Windows remarkably increases stability at high beam currents and at high annealing temperatures. With a minimal background signal, elemental analyses of samples containing nitrogen and/or carbon is possible by EDX and EELS. Nanoporous with pores from 10-60 nm.
Dimensions: (2) 100x1500 µm
Thickness: 15nm
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.